Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STXBP5L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308330.1 | NP_001295259.1 | 22 | 2656 | ||
NM_014980.2 | NP_055795.1 | 23 | 2728 | |||
XM_006713825.3 | XP_006713888.1 | 23 | 2898 | |||
XM_011513328.2 | XP_011511630.1 | 23 | 2703 | |||
XM_011513332.2 | XP_011511634.1 | 9 | 1205 | |||
XM_017007530.1 | XP_016863019.1 | 23 | 2659 | |||
XM_017007531.1 | XP_016863020.1 | 22 | 2641 | |||
XM_017007532.1 | XP_016863021.1 | 22 | 2590 | |||
XM_017007533.1 | XP_016863022.1 | 20 | 2316 | |||
GenBank mRNA | AB023223.1 | - | 23 | 2695 | ||
AK296544.1 | - | 22 | 2654 | |||
BC037531.2 | - | 11 | 1290 |