Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STXBP5L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308330.1 | NP_001295259.1 | 25 | 3270 | ||
NM_014980.2 | NP_055795.1 | 26 | 3342 | |||
XM_006713825.3 | XP_006713888.1 | 26 | 3512 | |||
XM_011513328.2 | XP_011511630.1 | 26 | 3317 | |||
XM_011513332.2 | XP_011511634.1 | 12 | 1819 | |||
XM_017007530.1 | XP_016863019.1 | 26 | 3273 | |||
XM_017007531.1 | XP_016863020.1 | 25 | 3255 | |||
XM_017007532.1 | XP_016863021.1 | 25 | 3204 | |||
XM_017007533.1 | XP_016863022.1 | 23 | 2930 | |||
GenBank mRNA | AB023223.1 | - | 26 | 3309 | ||
AK296544.1 | - | 25 | 3268 | |||
BC037531.2 | - | 14 | 1904 |