Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STXBP5L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308330.1 | NP_001295259.1 | 8 | 841 | ||
NM_014980.2 | NP_055795.1 | 8 | 841 | |||
XM_006713825.3 | XP_006713888.1 | 8 | 1011 | |||
XM_011513328.2 | XP_011511630.1 | 7 | 688 | |||
XM_011513331.2 | XP_011511633.1 | 8 | 838 | |||
XM_011513333.2 | XP_011511635.1 | 8 | 838 | |||
XM_017007530.1 | XP_016863019.1 | 8 | 772 | |||
XM_017007531.1 | XP_016863020.1 | 8 | 826 | |||
XM_017007532.1 | XP_016863021.1 | 8 | 775 | |||
XM_017007533.1 | XP_016863022.1 | 5 | 429 | |||
XM_017007534.1 | XP_016863023.1 | 8 | 838 | |||
XM_017007535.1 | XP_016863024.1 | 8 | 1029 | |||
GenBank mRNA | AB023223.1 | - | 8 | 808 | ||
AK296544.1 | - | 8 | 839 | |||
BC022029.1 | - | 8 | 819 |