Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TECR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138501.5 | NP_612510.1 | 1, 2 | 151 | ||
NR_038103.1 | 1, 2 | 151 | ||||
NR_038104.1 | 1, 2 | 151 | ||||
GenBank mRNA | AF038958.1 | - | 1, 2 | 97 | ||
AF038959.1 | - | 1, 2 | 33 | |||
AF222742.1 | - | 1, 2 | 72 | |||
AK001416.1 | - | 1, 2 | 115 | |||
AK315412.1 | - | 1, 2 | 115 | |||
BC000174.2 | - | 11 | ||||
BC002425.1 | - | 1, 2 | 83 | 3163592 | ||
BC005952.1 | - | 1, 2 | 94 | 4249841 | ||
BC007801.2 | - | 1, 2 | 101 | 4128702 | ||
BC013881.2 | - | 1, 2 | 99 | 3928698 | ||
BG705196.1 | - | 1, 2 | 154 | |||
BT007179.1 | - | 1 | 21 | |||
CR456892.1 | - | 1 | 21 |
Gene Symbol: GPBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC070132.1 | - | 115 |