Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TECR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321170.1 | NP_001308099.1 | 4 | 302 | ||
NM_138501.5 | NP_612510.1 | 3 | 228 | |||
NR_038103.1 | 3 | 260 | ||||
NR_038104.1 | 3 | 293 | ||||
XM_006722945.1 | XP_006723008.1 | 4 | 346 | |||
XM_011528442.2 | XP_011526744.1 | 3 | 176 | |||
GenBank mRNA | AF038958.1 | - | 3 | 174 | ||
AF222742.1 | - | 3 | 149 | |||
AK001416.1 | - | 3 | 192 | |||
AK094087.1 | - | 4 | 269 | |||
AK299134.1 | - | 4 | 266 | |||
AK315412.1 | - | 3 | 192 | |||
BC000174.2 | - | 2 | 88 | |||
BC002425.1 | - | 3 | 160 | 3163592 | ||
BC005952.1 | - | 3 | 171 | 4249841 | ||
BC007801.2 | - | 3 | 178 | 4128702 | ||
BC013881.2 | - | 3 | 176 | 3928698 | ||
BG705196.1 | - | 3 | 231 | |||
BT007179.1 | - | 2 | 98 | |||
CR456892.1 | - | 2 | 98 | |||
DB491783.1 | - | 3 | 236 |