Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA0391 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256679.1 | NP_001243608.1 | 2, 3 | 1040 | ||
NM_014672.3 | NP_055487.2 | 2, 3 | 1593 | |||
XM_005268237.3 | XP_005268294.1 | 2, 3 | 1492 | |||
XM_011537409.2 | XP_011535711.1 | 2, 3 | 1419 | |||
XM_011537410.2 | XP_011535712.1 | 2, 3 | 1477 | |||
XM_017021835.1 | XP_016877324.1 | 2, 3 | 1365 | |||
GenBank mRNA | AK301931.1 | - | 2, 3 | 814 | ||
BC032221.1 | - | 2, 3 | 1344 | 5206545 | ||
BC044580.1 | - | 2, 3 | 1362 | 4822711 | ||
BP373829.1 | - | 1, 2 | 448 | |||
BX161394.1 | - | 2, 3 | 1325 |