Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP6NL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080491.3 | NP_001073960.1 | 5 | 313 | ||
NM_014688.3 | NP_055503.1 | 6 | 644 | |||
XM_006717542.3 | XP_006717605.1 | 7 | 470 | |||
XM_011519762.2 | XP_011518064.1 | 8 | 803 | |||
XM_017016971.1 | XP_016872460.1 | 7 | 477 | |||
XM_017016972.1 | XP_016872461.1 | 6 | 436 | |||
XM_017016973.1 | XP_016872462.1 | 5 | 3640 | |||
GenBank mRNA | AB449916.1 | - | 4 | 214 | ||
AK292944.1 | - | 6 | 434 | |||
BC042943.2 | - | 6 | 482 | 5296060 | ||
CF121539.1 | - | 5 | 320 | |||
D13644.1 | - | 6 | 493 |