Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM131B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031690.2 | NP_001026860.2 | 4 | 345 | ||
NM_014690.4 | NP_055505.3 | 4 | 238 | |||
XM_005250073.3 | XP_005250130.1 | 4 | 559 | |||
XM_006716186.2 | XP_006716249.1 | 4 | 337 | |||
GenBank mRNA | AB018316.1 | - | 3 | 133 | ||
AK124557.1 | - | 4 | 293 | |||
AK291470.1 | - | 4 | 210 | |||
AL832579.2 | - | 3 | 1826 | |||
BC045611.1 | - | 4 | 350 | 5300056 | ||
BC050543.1 | - | 4 | 213 | 5768309 | ||
DA226407.1 | - | 4 | 337 |