Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080496.2 | NP_001073965.2 | 5 | 571 | ||
GenBank mRNA | AK289566.1 | - | 5 | 564 | ||
BC001725.1 | - | 5 | 499 | 3533857 | ||
D87447.1 | - | 5 | 508 |