Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TESPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098815.2 | NP_001092285.1 | 6, 7 | 468 | ||
NM_001136030.2 | NP_001129502.1 | 6, 7 | 565 | |||
NM_001261844.1 | NP_001248773.1 | 5, 6 | 317 | |||
NM_014796.2 | NP_055611.1 | 4, 5 | 408 | |||
XM_005269247.2 | XP_005269304.1 | 5, 6 | 391 | |||
XM_006719715.3 | XP_006719778.1 | 8, 9 | 899 | |||
XM_011539035.2 | XP_011537337.1 | 8, 9 | 912 | |||
XM_011539036.1 | XP_011537338.1 | 7, 8 | 733 | |||
XM_011539037.2 | XP_011537339.1 | 6, 7 | 4953 | |||
XM_017020262.1 | XP_016875751.1 | 8, 9 | 1962 | |||
XM_017020263.1 | XP_016875752.1 | 8, 9 | 1868 | |||
GenBank mRNA | AB018291.1 | - | 4, 5 | 606 | ||
AK295026.1 | - | 4, 5 | 259 | |||
AK298405.1 | - | 4, 5 | 234 | |||
AK303223.1 | - | 6, 7 | 491 | |||
AK315972.1 | - | 5, 6 | 271 | |||
BC044578.1 | - | 4, 5 | 238 | 5275264 | ||
BC132732.1 | - | 6, 7 | 468 | 40146754 | ||
BC132734.1 | - | 6, 7 | 468 | 40146756 |