Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286658.2 | NP_001273587.1 | 10 | 2357 | ||
NM_001286659.2 | NP_001273588.1 | 10 | 2357 | |||
NM_014832.4 | NP_055647.2 | 10 | 2357 | |||
XM_005266603.1 | XP_005266660.1 | 9 | 2760 | |||
XM_005266605.2 | XP_005266662.1 | 10 | 1685 | |||
XM_006719903.3 | XP_006719966.1 | 10 | 2045 | |||
XM_011535331.1 | XP_011533633.1 | 9 | 2724 | |||
XM_017020882.1 | XP_016876371.1 | 10 | 1566 | |||
XM_017020883.1 | XP_016876372.1 | 9 | 1560 | |||
GenBank mRNA | AB011175.1 | - | 10 | 2303 | ||
AB449885.1 | - | 10 | 1956 | |||
AK307493.1 | - | 10 | 2357 | |||
AY211910.1 | - | 1 | 116 | |||
BC151239.1 | - | 10 | 2295 | 8860512 | ||
CR749622.1 | - | 1 | 161 | |||
FM207106.1 | - | 10 | 1956 | |||
FM207107.1 | - | 10 | 1956 |