Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRGAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033117.2 | NP_001028289.1 | 13 | 2246 | ||
NM_014850.3 | NP_055665.1 | 13 | 2318 | |||
XM_011534295.1 | XP_011532597.1 | 14 | 2333 | |||
XM_011534296.1 | XP_011532598.1 | 15 | 2450 | |||
XM_011534301.2 | XP_011532603.1 | 11 | 4162 | |||
XM_011534303.1 | XP_011532605.1 | 9 | 1058 | |||
XM_017007574.1 | XP_016863063.1 | 13 | 2656 | |||
XM_017007575.1 | XP_016863064.1 | 13 | 2584 | |||
XM_017007576.1 | XP_016863065.1 | 15 | 1825 | |||
XM_017007577.1 | XP_016863066.1 | 15 | 1764 | |||
XM_017007578.1 | XP_016863067.1 | 13 | 2684 | |||
XM_017007579.1 | XP_016863068.1 | 13 | 2652 | |||
XM_017007580.1 | XP_016863069.1 | 15 | 1813 | |||
GenBank mRNA | AB007871.2 | - | 13 | 1470 | ||
AF427144.1 | - | 13 | 1686 | |||
AF464189.1 | - | 13 | 1774 | |||
BC039300.1 | - | 15 | 1893 |