Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRGAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033117.2 | NP_001028289.1 | 3 | 1020 | ||
NM_014850.3 | NP_055665.1 | 3 | 1020 | |||
XM_011534295.1 | XP_011532597.1 | 3 | 1020 | |||
XM_011534296.1 | XP_011532598.1 | 4 | 1137 | |||
XM_017007574.1 | XP_016863063.1 | 3 | 1358 | |||
XM_017007575.1 | XP_016863064.1 | 3 | 1358 | |||
XM_017007576.1 | XP_016863065.1 | 5 | 527 | |||
XM_017007577.1 | XP_016863066.1 | 5 | 466 | |||
XM_017007578.1 | XP_016863067.1 | 3 | 1386 | |||
XM_017007579.1 | XP_016863068.1 | 3 | 1354 | |||
XM_017007580.1 | XP_016863069.1 | 5 | 587 | |||
GenBank mRNA | AB007871.2 | - | 3 | 244 | ||
AF427144.1 | - | 3 | 388 | |||
AF464189.1 | - | 3 | 548 | |||
AK094355.1 | - | 3 | 326 | |||
BC039300.1 | - | 3 | 355 |