Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KBTBD11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014867.2 | NP_055682.1 | 2 | 2793 | ||
XM_011534771.2 | XP_011533073.1 | 2 | 3463 | |||
XM_011534772.2 | XP_011533074.1 | 3 | 2862 | |||
XM_017014114.1 | XP_016869603.1 | 4 | 3397 | |||
XM_017014115.1 | XP_016869604.1 | 3 | 3251 | |||
XM_017014116.1 | XP_016869605.1 | 3 | 2918 | |||
XM_017014117.1 | XP_016869606.1 | 2 | 3182 | |||
GenBank mRNA | AB018254.1 | - | 2 | 2790 | ||
AY762229.1 | - | 1 | 1827 |