Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDAC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015053.1 | NP_001015053.1 | 4 | 640 | ||
NM_005474.4 | NP_005465.2 | 4 | 637 | |||
XM_005256905.1 | XP_005256962.1 | 4 | 638 | |||
XM_005256906.4 | XP_005256963.1 | 4 | 596 | |||
XM_011524149.1 | XP_011522451.1 | 4 | 638 | |||
XM_011524150.1 | XP_011522452.1 | 3 | 387 | |||
XM_017023988.1 | XP_016879477.1 | 6 | 749 | |||
XM_017023989.1 | XP_016879478.1 | 6 | 715 | |||
XM_017023990.1 | XP_016879479.1 | 6 | 740 | |||
XM_017023991.1 | XP_016879480.1 | 6 | 749 | |||
XM_017023992.1 | XP_016879481.1 | 6 | 750 | |||
XM_017023993.1 | XP_016879482.1 | 6 | 749 | |||
XM_017023994.1 | XP_016879483.1 | 2 | 730 | |||
XM_017023995.1 | XP_016879484.1 | 4 | 632 | |||
XM_017023996.1 | XP_016879485.1 | 5 | 812 | |||
GenBank mRNA | AB011172.2 | - | 4 | 609 | ||
AF132608.1 | - | 3 | 305 | |||
AK294760.1 | - | 4 | 605 | |||
BC013140.1 | - | 1 | 87 | |||
BC051824.1 | - | 4 | 516 | 6043491 | ||
BM564861.1 | - | 4 | 635 |