Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCL2L10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306168.1 | NP_001293097.1 | 1 | 223 | ||
NM_020396.3 | NP_065129.1 | 1 | 223 | |||
GenBank mRNA | AF285092.1 | - | 1 | 174 | ||
AF326964.1 | - | 1 | 223 | |||
BC093826.1 | - | 1 | 216 | 7939671 | ||
BC093828.1 | - | 1 | 216 | 7939673 | ||
BC104442.1 | - | 1 | 217 | 40022820 | ||
BC104443.1 | - | 1 | 217 | 40022822 | ||
BC143227.1 | - | 1 | 216 |