Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256670.1 | NP_001243599.1 | 16, 17 | 2420 | ||
NM_005700.4 | NP_005691.2 | 17, 18 | 2510 | |||
NM_130443.3 | NP_569710.2 | 17, 18 | 2513 | |||
GenBank mRNA | AB017970.1 | - | 11, 12 | 1320 | ||
AJ271216.1 | - | 17, 18 | 2109 | |||
AK021449.1 | - | 7, 8 | 828 | |||
AK025984.1 | - | 13, 14 | 1518 | |||
AK222847.1 | - | 17, 18 | 2088 | |||
AK297199.1 | - | 16, 17 | 1995 | |||
AK304581.1 | - | 18, 19 | 2011 | |||
AK315478.1 | - | 17, 18 | 2501 | |||
AL137670.1 | - | 1, 2 | 55 | |||
AL833475.1 | - | 17, 18 | 2115 | |||
BC001446.2 | - | 16, 17 | 2073 | 3138626 | ||
BC007221.1 | - | 17, 18 | 2099 | 3349135 | ||
BC014038.2 | - | 17, 18 | 2073 | 4547232 | ||
BC024271.1 | - | 16, 17 | 2069 | 4564651 | ||
CD366941.1 | - | 1 | 585 |