Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HUWE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031407.6 | NP_113584.3 | 43 | 6043 | ||
XM_005261965.3 | XP_005262022.1 | 42 | 6110 | |||
XM_011530747.1 | XP_011529049.1 | 44 | 6286 | |||
XM_011530751.2 | XP_011529053.1 | 43 | 6154 | |||
XM_017029191.1 | XP_016884680.1 | 42 | 9038 | |||
XM_017029192.1 | XP_016884681.1 | 42 | 9038 | |||
XM_017029193.1 | XP_016884682.1 | 42 | 9017 | |||
XM_017029194.1 | XP_016884683.1 | 42 | 9038 | |||
XM_017029195.1 | XP_016884684.1 | 42 | 9038 | |||
XM_017029196.1 | XP_016884685.1 | 42 | 9038 | |||
XM_017029197.1 | XP_016884686.1 | 42 | 9038 | |||
XM_017029198.1 | XP_016884687.1 | 43 | 6380 | |||
XM_017029199.1 | XP_016884688.1 | 44 | 6307 | |||
XM_017029200.1 | XP_016884689.1 | 43 | 6177 | |||
XM_017029201.1 | XP_016884690.1 | 43 | 6194 | |||
XM_017029202.1 | XP_016884691.1 | 43 | 6170 | |||
XM_017029203.1 | XP_016884692.1 | 43 | 6176 | |||
XM_017029204.1 | XP_016884693.1 | 41 | 8788 | |||
XM_017029205.1 | XP_016884694.1 | 41 | 8768 | |||
XM_017029206.1 | XP_016884695.1 | 42 | 9038 | |||
GenBank mRNA | AB002310.3 | - | 12 | 2145 | ||
AB071605.1 | - | 16 | 2743 | |||
AY772009.1 | - | 41 | 5688 | |||
AY929612.1 | - | 40 | 5640 | |||
DQ097177.1 | - | 43 | 6042 |