Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139022.2 | NP_620591.3 | 9 | 933 | ||
XM_005252719.4 | XP_005252776.1 | 9 | 927 | |||
XM_005252720.4 | XP_005252777.1 | 7 | 2226 | |||
XM_011519815.2 | XP_011518117.2 | 8 | 849 | |||
XM_011519818.2 | XP_011518120.2 | 8 | 849 | |||
XM_011519819.1 | XP_011518121.1 | 9 | 815 | |||
XM_011519829.2 | XP_011518131.2 | 7 | 1816 | |||
XM_017017064.1 | XP_016872553.1 | 8 | 1399 | |||
XM_017017065.1 | XP_016872554.1 | 8 | 1399 | |||
XM_017017066.1 | XP_016872555.1 | 7 | 1315 | |||
XM_017017067.1 | XP_016872556.1 | 9 | 819 | |||
XM_017017069.1 | XP_016872558.1 | 6 | 2698 | |||
XM_017017071.1 | XP_016872560.1 | 6 | 2291 | |||
GenBank mRNA | AF125569.1 | - | 8 | 851 | ||
AF176070.1 | - | 8 | 817 | |||
AK128812.1 | - | 8 | 2897 | |||
AY039001.1 | - | 9 | 802 | |||
AY303780.1 | - | 7 | 631 | |||
BC016693.1 | - | 9 | 881 | 4071111 |