Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP9A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006045.2 | NP_006036.1 | 24, 25 | 2677 | ||
XM_011528481.2 | XP_011526783.1 | 24, 25 | 2705 | |||
GenBank mRNA | AB014511.1 | - | 19, 20 | 2231 | ||
AK172802.1 | - | 22, 23 | 2366 | |||
AK299064.1 | - | 24, 25 | 2606 | |||
BC110591.1 | - | 20, 21 | 2169 | |||
BC110592.1 | - | 20, 21 | 2169 |