Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP9A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006045.2 | NP_006036.1 | 10 | 892 | ||
XM_011528481.2 | XP_011526783.1 | 10 | 920 | |||
GenBank mRNA | AB014511.1 | - | 5 | 446 | ||
AK172802.1 | - | 8 | 581 | |||
AK299064.1 | - | 10 | 821 | |||
BC110591.1 | - | 6 | 384 | |||
BC110592.1 | - | 6 | 384 | |||
BF089115.1 | - | 2 | 157 |