Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARPC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278556.1 | NP_001265485.1 | 7 | 903 | ||
NM_001287222.1 | NP_001274151.1 | 7 | 900 | |||
GenBank mRNA | AF006086.1 | - | 7 | 777 | ||
AI144468.1 | - | 1 | 56 | |||
AK297309.1 | - | 7 | 572 | |||
BC067747.1 | - | 7 | 809 | 5302338 | ||
BC078162.1 | - | 7 | 786 | 6148231 | ||
BM999380.1 | - | 1 | 53 | |||
DQ328220.1 | - | 7 | 833 |