Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL4C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282431.1 | NP_001269360.1 | 1 | 598 | ||
NM_005737.3 | NP_005728.2 | 1 | 594 | |||
GenBank mRNA | AB016811.1 | - | 1 | 102 | ||
AF493892.1 | - | 1 | 131 | |||
AJ579850.1 | - | 1 | 145 | |||
AJ579851.1 | - | 1 | 145 | |||
AK314119.1 | - | 1 | 135 | |||
BC089043.1 | - | 1 | 598 | 6578402 | ||
BT019378.1 | - | 1 | 131 | |||
BT019379.1 | - | 1 | 131 | |||
Y17804.1 | - | 1 | 145 |