Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASGRP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098670.1 | NP_001092140.1 | 9 | 1099 | ||
NM_001098671.1 | NP_001092141.1 | 9 | 1139 | |||
NM_001318398.1 | NP_001305327.1 | 9 | 1059 | |||
NM_153819.1 | NP_722541.1 | 9 | 1165 | |||
XM_005273707.3 | XP_005273764.3 | 9 | 1490 | |||
XM_011544718.1 | XP_011543020.1 | 9 | 1490 | |||
XM_011544720.1 | XP_011543022.1 | 9 | 1570 | |||
XM_011544721.1 | XP_011543023.1 | 9 | 1102 | |||
XM_011544722.1 | XP_011543024.1 | 9 | 1155 | |||
XM_011544723.2 | XP_011543025.1 | 9 | 1104 | |||
XM_011544725.2 | XP_011543027.1 | 8 | 936 | |||
XM_017017082.1 | XP_016872571.1 | 8 | 2140 | |||
XM_017017083.1 | XP_016872572.1 | 8 | 2140 | |||
XM_017017084.1 | XP_016872573.1 | 9 | 1133 | |||
XM_017017085.1 | XP_016872574.1 | 6 | 923 | |||
XM_017017086.1 | XP_016872575.1 | 6 | 639 | |||
GenBank mRNA | AF043722.1 | - | 10 | 1271 | ||
AF043723.1 | - | 9 | 1165 | |||
AF081194.1 | - | 9 | 1072 | |||
AK092852.1 | - | 4 | 1261 | |||
AK092882.1 | - | 8 | 1835 | |||
AK122694.1 | - | 9 | 1023 | |||
AK314637.1 | - | 8 | 1050 | |||
BC110306.1 | - | 9 | 1073 | |||
BC117151.1 | - | 9 | 1163 | 40125702 | ||
U78170.1 | - | 9 | 1072 | |||
Y12336.1 | - | 9 | 1165 |