Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASGRP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098670.1 | NP_001092140.1 | 5 | 535 | ||
NM_001098671.1 | NP_001092141.1 | 5 | 575 | |||
NM_001318398.1 | NP_001305327.1 | 5 | 495 | |||
NM_153819.1 | NP_722541.1 | 5 | 601 | |||
XM_005273707.3 | XP_005273764.3 | 5 | 926 | |||
XM_011544718.1 | XP_011543020.1 | 5 | 926 | |||
XM_011544720.1 | XP_011543022.1 | 5 | 1006 | |||
XM_011544721.1 | XP_011543023.1 | 5 | 538 | |||
XM_011544722.1 | XP_011543024.1 | 5 | 591 | |||
XM_011544723.2 | XP_011543025.1 | 5 | 540 | |||
XM_011544725.2 | XP_011543027.1 | 3 | 290 | |||
XM_017017082.1 | XP_016872571.1 | 4 | 1576 | |||
XM_017017083.1 | XP_016872572.1 | 4 | 1576 | |||
XM_017017084.1 | XP_016872573.1 | 5 | 569 | |||
XM_017017085.1 | XP_016872574.1 | 2 | 359 | |||
GenBank mRNA | AF043722.1 | - | 6 | 707 | ||
AF043723.1 | - | 5 | 601 | |||
AF081194.1 | - | 5 | 508 | |||
AK092882.1 | - | 4 | 1271 | |||
AK122694.1 | - | 5 | 459 | |||
AK124023.1 | - | 4 | 2071 | |||
AK314637.1 | - | 4 | 486 | |||
BC110306.1 | - | 5 | 509 | |||
BC117151.1 | - | 5 | 599 | 40125702 | ||
DA529151.1 | - | 5 | 575 | |||
U78170.1 | - | 5 | 508 | |||
Y12336.1 | - | 5 | 601 |