Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021147.4 | NP_066970.3 | 2 | 656 | ||
NR_125346.1 | 2 | 1032 | ||||
NR_125347.1 | 2 | 661 | ||||
NR_125348.1 | 1 | 526 | ||||
GenBank mRNA | AK026075.1 | - | 2 | 619 | ||
BC004877.2 | - | 2 | 912 | |||
BC143907.1 | - | 2 | 187 | |||
BX096363.1 | - | 1 | 194 | |||
HY011414.1 | - | 2 | 318 | |||
X52486.1 | - | 1 | 427 |