Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCIRG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006019.3 | NP_006010.2 | 11, 12 | 1433 | ||
NM_006053.3 | NP_006044.1 | 6, 7 | 1192 | |||
XM_005273709.3 | XP_005273766.1 | 12, 13 | 1592 | |||
XM_011544726.2 | XP_011543028.1 | 11, 12 | 1332 | |||
XM_017017089.1 | XP_016872578.1 | 11, 12 | 1428 | |||
XM_017017090.1 | XP_016872579.1 | 10, 11 | 1771 | |||
GenBank mRNA | AF025374.1 | - | 6, 7 | 1192 | ||
BC018133.1 | - | 11, 12 | 1412 | 3905446 | ||
BC032465.1 | - | 11, 12 | 1405 | 5210733 | ||
U45285.1 | - | 11, 12 | 1361 |