Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RTN3 SIRNA Assays ›
Gene Symbol:
RTN3
Gene Name:
reticulon 3
Gene Aliases:
ASYIP, HAP, NSPL2, NSPLII, RTN3-A1
Chromosome Location:
Chr.11: 63681315 - 63759891 on Build GRCh38
UniGene ID:
Hs.743229
Species:
Human
siRNA ID
HSS145573
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RTN3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001265589.1
NP_001252518.1
4
2747
NM_001265590.1
NP_001252519.1
3
2411
NM_001265591.1
NP_001252520.1
2
359
NM_006054.3
NP_006045.1
2
359
NM_201428.2
NP_958831.1
3
2690
NM_201429.2
NP_958832.1
3
416
NM_201430.2
NP_958833.1
2
359
NR_049750.1
2
359
NR_049751.1
2
359
XM_011544730.2
XP_011543032.1
4
2837
XM_011544731.2
XP_011543033.1
3
551
GenBank mRNA
AB209771.1
-
2
296
AF059524.1
-
2
296
AF119297.1
-
2
308
AK075412.1
-
2
309
AK094965.1
-
2
1412
AK122727.1
-
2
812
AK127079.1
-
3
2386
AK222898.1
-
2
296
AK295361.1
-
3
321
AK295534.1
-
2
213
AK295758.1
-
3
2339
AK297529.1
-
2
236
AK297533.1
-
2
309
AK304100.1
-
4
2697
AK307811.1
-
3
2636
AK308083.1
-
2
304
AK309766.1
-
3
2365
AY427821.2
-
3
2657
AY750848.1
-
4
2714
BC000634.1
-
2
267
BC010556.1
-
2
272
BC011394.1
-
2
308
BC022993.1
-
2
298
BC100822.2
-
2
289
BC100823.2
-
2
289
BC105981.1
-
2
240
BC105982.1
-
2
271
BC118550.1
-
2
289
BC118628.1
-
2
289
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
apoptotic process
chemical synaptic transmission
viral process
vesicle-mediated transport
endoplasmic reticulum tubular network organization
endoplasmic reticulum tubular network assembly
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-x88h5:80/100.66.78.96:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline