Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001265589.1 | NP_001252518.1 | 5 | 3019 | ||
NM_001265590.1 | NP_001252519.1 | 4 | 2683 | |||
NM_006054.3 | NP_006045.1 | 3 | 631 | |||
NM_201428.2 | NP_958831.1 | 4 | 2962 | |||
NM_201429.2 | NP_958832.1 | 4 | 688 | |||
NM_201430.2 | NP_958833.1 | 3 | 631 | |||
XM_011544730.2 | XP_011543032.1 | 5 | 3109 | |||
XM_011544731.2 | XP_011543033.1 | 4 | 823 | |||
GenBank mRNA | AB209771.1 | - | 3 | 568 | ||
AF119297.1 | - | 3 | 580 | |||
AK075412.1 | - | 3 | 581 | |||
AK094965.1 | - | 3 | 1684 | |||
AK122727.1 | - | 3 | 1084 | |||
AK127079.1 | - | 4 | 2658 | |||
AK222898.1 | - | 3 | 568 | |||
AK295361.1 | - | 4 | 593 | |||
AK295534.1 | - | 3 | 485 | |||
AK295758.1 | - | 4 | 2611 | |||
AK304100.1 | - | 5 | 2969 | |||
AK307811.1 | - | 4 | 2908 | |||
AK308083.1 | - | 3 | 576 | |||
AK309766.1 | - | 4 | 2637 | |||
AY427821.2 | - | 4 | 2929 | |||
AY750848.1 | - | 5 | 2986 | |||
BC000634.1 | - | 3 | 539 | |||
BC010556.1 | - | 3 | 544 | |||
BC011394.1 | - | 3 | 580 | |||
BC022993.1 | - | 3 | 570 | |||
BC100822.2 | - | 3 | 561 | |||
BC100823.2 | - | 3 | 561 | |||
BC105981.1 | - | 3 | 512 | |||
BC105982.1 | - | 3 | 543 | |||
BC118550.1 | - | 3 | 561 | |||
BC118628.1 | - | 3 | 561 |