Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267551.1 | NP_001254480.1 | 6 | 642 | ||
NM_001267552.1 | NP_001254481.1 | 7 | 739 | |||
NM_007169.2 | NP_009100.2 | 6 | 700 | |||
NM_148172.2 | NP_680477.1 | 6 | 708 | |||
NM_148173.1 | NP_680478.1 | 6 | 587 | |||
XM_006721418.3 | XP_006721481.2 | 6 | 589 | |||
XM_017024016.1 | XP_016879505.1 | 4 | 495 | |||
GenBank mRNA | AF044214.1 | - | 6 | 673 | ||
AF113126.1 | - | 6 | 677 | |||
BC050593.1 | - | 6 | 685 | 6148983 |