Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDIPT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286585.1 | NP_001273514.1 | 2 | 527 | ||
NM_001286586.1 | NP_001273515.1 | 3 | 577 | |||
NM_006319.4 | NP_006310.1 | 3 | 662 | |||
XM_005255038.4 | XP_005255095.1 | 1 | 549 | |||
GenBank mRNA | AF014807.1 | - | 3 | 578 | ||
AK094468.1 | - | 2 | 496 | |||
AK097691.1 | - | 3 | 1108 | |||
AK131227.1 | - | 1 | 455 | |||
AK131349.1 | - | 3 | 568 | |||
AK222676.1 | - | 3 | 541 | |||
AK290632.1 | - | 4 | 436 | |||
AK310988.1 | - | 562 | ||||
AK314544.1 | - | 3 | 593 | |||
BC001444.2 | - | 3 | 498 | 3139134 | ||
BI490060.1 | - | 2 | 490 | |||
BT007301.1 | - | 3 | 231 | |||
BX333193.2 | - | 3 | 533 | |||
CR542016.1 | - | 3 | 231 |