Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ARIH2 SIRNA Assays ›
Gene Symbol:
ARIH2
Gene Name:
ariadne RBR E3 ubiquitin protein ligase 2
Gene Aliases:
ARI2, TRIAD1
Chromosome Location:
Chr.3: 48918820 - 48985541 on Build GRCh38
UniGene ID:
Hs.633601
Species:
Human
siRNA ID
HSS145625
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ARIH2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001317333.1
NP_001304262.1
7
927
NM_001317334.1
NP_001304263.1
4
389
NM_006321.3
NP_006312.1
6
805
XM_011533265.1
XP_011531567.1
6
793
XM_011533266.1
XP_011531568.1
7
915
XM_011533267.1
XP_011531569.1
6
847
XM_011533268.1
XP_011531570.1
5
740
XM_011533270.1
XP_011531572.1
7
827
XM_011533271.1
XP_011531573.1
6
477
XM_011533273.1
XP_011531575.1
8
858
XM_011533275.2
XP_011531577.1
6
747
XM_017005531.1
XP_016861020.1
6
847
XM_017005532.1
XP_016861021.1
5
740
XM_017005533.1
XP_016861022.1
5
1187
XM_017005534.1
XP_016861023.1
5
396
XM_017005535.1
XP_016861024.1
5
604
XM_017005536.1
XP_016861025.1
8
961
XM_017005537.1
XP_016861026.1
7
827
XM_017005538.1
XP_016861027.1
7
873
XM_017005539.1
XP_016861028.1
6
477
XM_017005540.1
XP_016861029.1
8
858
XM_017005541.1
XP_016861030.1
8
928
GenBank mRNA
AF099149.1
-
5
609
AF183427.1
-
5
608
AJ130978.1
-
5
609
AK001800.1
-
4
489
AK091240.1
-
5
1352
AK223550.1
-
7
911
AK307694.1
-
3
201
AK313434.1
-
4
469
BC000422.2
-
6
765
2964505
BM928477.1
-
4
336
CR456785.1
-
4
465
Back To Top
More Information
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Function(s)
Process(es)
protein polyubiquitination
multicellular organism development
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
developmental cell growth
protein K63-linked ubiquitination
protein K48-linked ubiquitination
hematopoietic stem cell proliferation
positive regulation of protein targeting to mitochondrion
ubiquitin-protein transferase activity
protein binding
zinc ion binding
ligase activity
ubiquitin conjugating enzyme binding
ubiquitin protein ligase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0