Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMM23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006327.3 | NP_006318.1 | 7 | 1178 | ||
NR_073029.1 | 8 | 1302 | ||||
NR_073030.1 | 6 | 1119 | ||||
GenBank mRNA | AF116628.2 | - | 1 | 50 | ||
AK223331.1 | - | 7 | 1118 | |||
AY442341.1 | - | 7 | 1129 | |||
BC062707.1 | - | 7 | 1179 | 4772587 | ||
BC066951.1 | - | 7 | 1128 | 5264814 | ||
BU619060.1 | - | 1 | 58 | |||
U38436.1 | - | 1 | 137 |