Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNHIT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006349.2 | NP_006340.1 | 3, 4 | 758 | ||
XM_011515739.2 | XP_011514041.1 | 3, 4 | 489 | |||
GenBank mRNA | AF093571.1 | - | 3, 4 | 336 | ||
AK125123.1 | - | 4, 5 | 3755 | |||
BC017333.2 | - | 3, 4 | 758 | 4647446 | ||
CR456992.1 | - | 3, 4 | 266 | |||
U61837.1 | - | 3, 4 | 296 |