Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIVA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006427.3 | NP_006418.2 | 3, 4 | 540 | ||
NM_021709.2 | NP_068355.1 | 2, 3 | 345 | |||
GenBank mRNA | AF033111.1 | - | 3, 4 | 366 | ||
AK223332.1 | - | 3, 4 | 526 | |||
BC034562.1 | - | 2, 3 | 319 | 3860364 | ||
BU195069.1 | - | 3, 4 | 542 |