Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIVA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006427.3 | NP_006418.2 | 4 | 581 | ||
NM_021709.2 | NP_068355.1 | 3 | 386 | |||
XM_011536360.2 | XP_011534662.1 | 4 | 1503 | |||
GenBank mRNA | AF033111.1 | - | 4 | 407 | ||
AF401214.1 | - | 1 | 24 | |||
AK124078.1 | - | 1316 | ||||
AK128704.1 | - | 2 | 3961 | |||
AK223332.1 | - | 4 | 567 | |||
BC034562.1 | - | 3 | 360 | 3860364 | ||
CK001877.1 | - | 4 | 575 |