Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASL10A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006477.4 | NP_006468.1 | 2 | 774 | ||
XM_011529821.1 | XP_011528123.1 | 3 | 492 | |||
XM_011529822.1 | XP_011528124.1 | 3 | 1248 | |||
XM_011529823.1 | XP_011528125.1 | 2 | 355 | |||
GenBank mRNA | BC022473.1 | - | 2 | 774 | 4794714 | |
BC040434.1 | - | 2 | 504 | 5727949 | ||
BC050739.1 | - | 2 | 710 | 5244128 | ||
BC058077.1 | - | 2 | 683 | 5240551 | ||
Y07847.1 | - | 2 | 585 |