Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCGB1D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006552.1 | NP_006543.1 | 2 | 149 | ||
GenBank mRNA | AJ224171.1 | - | 2 | 149 | ||
BC062693.1 | - | 2 | 179 | 30327780 | ||
BC069170.1 | - | 2 | 149 | 7216920 | ||
BC069289.1 | - | 2 | 149 | 7262693 |