Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DMRT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130865.2 | NP_001124337.1 | 6 | 1825 | ||
NM_006557.6 | NP_006548.1 | 4 | 2386 | |||
NM_181872.4 | NP_870987.2 | 4 | 1618 | |||
XM_011517687.1 | XP_011515989.1 | 6 | 4166 | |||
XM_011517690.2 | XP_011515992.1 | 5 | 2246 | |||
XM_011517694.2 | XP_011515996.1 | 3 | 1777 | |||
XM_017014213.1 | XP_016869702.1 | 6 | 2568 | |||
XM_017014214.1 | XP_016869703.1 | 5 | 2243 | |||
XM_017014215.1 | XP_016869704.1 | 4 | 2087 | |||
GenBank mRNA | AF284223.1 | - | 3 | 1913 | ||
AF284224.1 | - | 4 | 1496 | |||
AF284225.1 | - | 3 | 1418 | |||
AK290794.1 | - | 6 | 2131 | |||
BC030204.1 | - | 6 | 1825 | |||
BC136493.1 | - | 3 | 1497 | 9020482 |