Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LBX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006562.4 | NP_006553.2 | 1 | 291 | ||
GenBank mRNA | BC069156.1 | - | 1 | 292 | 7217017 | |
BC136321.1 | - | 1 | 263 | 9020308 |