Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006578.3 | NP_006569.1 | 9 | 903 | ||
NM_016194.3 | NP_057278.2 | 11 | 1008 | |||
XM_011521162.2 | XP_011519464.1 | 9 | 1260 | |||
XM_011521163.2 | XP_011519465.1 | 9 | 785 | |||
XM_017021867.1 | XP_016877356.1 | 6 | 2526 | |||
GenBank mRNA | AF017656.1 | - | 9 | 916 | ||
AF300650.1 | - | 10 | 943 | |||
AF501885.1 | - | 9 | 817 | |||
AK094776.1 | - | 2 | 631 | |||
AK125702.1 | - | 6 | 1424 | |||
AK290330.1 | - | 9 | 911 | |||
AK314775.1 | - | 9 | 861 | |||
AL117471.1 | - | 7 | 615 | |||
BC013997.2 | - | 9 | 900 | 3621195 |