Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006578.3 | NP_006569.1 | 9 | 952 | ||
NM_016194.3 | NP_057278.2 | 11 | 1057 | |||
XM_011521162.2 | XP_011519464.1 | 9 | 1309 | |||
XM_011521163.2 | XP_011519465.1 | 9 | 834 | |||
XM_017021867.1 | XP_016877356.1 | 6 | 2575 | |||
GenBank mRNA | AF017656.1 | - | 9 | 965 | ||
AF300650.1 | - | 10 | 992 | |||
AF501885.1 | - | 9 | 866 | |||
AK094776.1 | - | 2 | 680 | |||
AK125702.1 | - | 6 | 1473 | |||
AK290330.1 | - | 9 | 960 | |||
AK314775.1 | - | 9 | 910 | |||
AL117471.1 | - | 7 | 664 | |||
BC013997.2 | - | 9 | 949 | 3621195 |