Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTGES3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282601.1 | NP_001269530.1 | 7 | 1267 | ||
NM_001282602.1 | NP_001269531.1 | 7 | 1240 | |||
NM_001282603.1 | NP_001269532.1 | 7 | 1231 | |||
NM_001282604.1 | NP_001269533.1 | 9 | 1379 | |||
NM_001282605.1 | NP_001269534.1 | 6 | 1177 | |||
NM_006601.6 | NP_006592.3 | 8 | 1330 | |||
NR_104219.1 | 8 | 2239 | ||||
XM_005268576.4 | XP_005268633.1 | 8 | 2920 | |||
XM_006719199.2 | XP_006719262.1 | 7 | 1589 | |||
XM_011537774.2 | XP_011536076.1 | 7 | 1616 | |||
GenBank mRNA | AK098214.1 | - | 8 | 2239 | ||
AK291945.1 | - | 8 | 1084 | |||
AK298160.1 | - | 7 | 1053 | |||
BC003005.1 | - | 8 | 1027 | 2821965 | ||
BC021167.1 | - | 4 | 581 |