Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ARPP21 SIRNA Assays ›
Gene Symbol:
ARPP21
Gene Name:
cAMP regulated phosphoprotein 21
Gene Aliases:
ARPP-21, R3HDM3, RCS, TARPP
Chromosome Location:
Chr.3: 35638981 - 35798933 on Build GRCh38
UniGene ID:
Hs.475902
Species:
Human
siRNA ID
HSS145792
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ARPP21
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001025068.1
NP_001020239.1
3
354
NM_001025069.1
NP_001020240.1
3
363
NM_001267616.1
NP_001254545.1
3
1510
NM_001267617.1
NP_001254546.1
3
320
NM_001267618.1
NP_001254547.1
2
451
NM_001267619.1
NP_001254548.1
2
285
NM_016300.4
NP_057384.2
3
337
NM_198399.1
NP_938409.1
2
345
XM_005264811.2
XP_005264868.1
3
366
XM_006712943.2
XP_006713006.1
3
366
XM_006712944.2
XP_006713007.1
3
366
XM_011533298.2
XP_011531600.1
3
366
XM_011533299.2
XP_011531601.1
3
910
XM_011533300.2
XP_011531602.1
3
529
XM_011533301.2
XP_011531603.1
3
360
XM_011533302.2
XP_011531604.1
4
452
XM_011533303.2
XP_011531605.1
4
1002
XM_017005574.1
XP_016861063.1
3
456
XM_017005575.1
XP_016861064.1
4
12089
XM_017005576.1
XP_016861065.1
3
910
XM_017005577.1
XP_016861066.1
3
360
XM_017005578.1
XP_016861067.1
3
336
XM_017005579.1
XP_016861068.1
3
910
XM_017005580.1
XP_016861069.1
4
1002
XM_017005581.1
XP_016861070.1
4
12089
XM_017005582.1
XP_016861071.1
3
910
XM_017005583.1
XP_016861072.1
3
360
XM_017005584.1
XP_016861073.1
3
910
XM_017005585.1
XP_016861074.1
3
910
XM_017005586.1
XP_016861075.1
3
366
XM_017005587.1
XP_016861076.1
3
529
XM_017005588.1
XP_016861077.1
4
1002
XM_017005589.1
XP_016861078.1
4
452
XM_017005590.1
XP_016861079.1
4
12089
XM_017005591.1
XP_016861080.1
3
910
XM_017005592.1
XP_016861081.1
3
910
XM_017005593.1
XP_016861082.1
3
456
XM_017005594.1
XP_016861083.1
3
360
XM_017005595.1
XP_016861084.1
4
1002
XM_017005596.1
XP_016861085.1
3
910
XM_017005597.1
XP_016861086.1
3
910
XM_017005598.1
XP_016861087.1
4
1002
XM_017005599.1
XP_016861088.1
3
910
XM_017005600.1
XP_016861089.1
3
529
XM_017005601.1
XP_016861090.1
3
360
XM_017005602.1
XP_016861091.1
3
910
XM_017005603.1
XP_016861092.1
3
529
XM_017005604.1
XP_016861093.1
3
360
XM_017005605.1
XP_016861094.1
4
1002
XM_017005606.1
XP_016861095.1
4
452
XM_017005607.1
XP_016861096.1
3
910
XM_017005608.1
XP_016861097.1
3
910
XM_017005609.1
XP_016861098.1
4
1002
XM_017005610.1
XP_016861099.1
3
910
XM_017005611.1
XP_016861100.1
3
910
XM_017005612.1
XP_016861101.1
3
910
GenBank mRNA
AF112220.1
-
2
310
AK055064.1
-
3
363
AK289649.1
-
2
300
AK289868.1
-
3
357
AK294484.1
-
2
298
AV718064.1
-
3
354
AV718166.1
-
3
363
BC017805.1
-
2
345
BC036399.1
-
3
325
5263894
BC041385.1
-
3
1098
BC051828.1
-
3
337
6012704
BC066651.1
-
1
864
BG535216.1
-
2
346
BX488342.1
-
2
449
DA275579.1
-
2
451
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
cellular response to heat
molecular_function
nucleic acid binding
calmodulin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0