Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF273 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021148.2 | NP_066971.2 | 4 | 435 | ||
NR_003099.1 | 5 | 524 | ||||
XM_011515746.2 | XP_011514048.1 | 3 | 370 | |||
XM_017011688.1 | XP_016867177.1 | 4 | 461 | |||
XM_017011689.1 | XP_016867178.1 | 9 | 6449 | |||
XM_017011690.1 | XP_016867179.1 | 8 | 6360 | |||
XM_017011691.1 | XP_016867180.1 | 9 | 3651 | |||
XM_017011692.1 | XP_016867181.1 | 7 | 5835 | |||
XM_017011693.1 | XP_016867182.1 | 8 | 1541 | |||
XM_017011694.1 | XP_016867183.1 | 8 | 899 | |||
XM_017011695.1 | XP_016867184.1 | 5 | 510 | |||
XM_017011696.1 | XP_016867185.1 | 6 | 736 | |||
XM_017011697.1 | XP_016867186.1 | 7 | 6271 | |||
XM_017011698.1 | XP_016867187.1 | 6 | 1388 | |||
GenBank mRNA | AK090648.1 | - | 8 | 1542 | ||
AL832810.1 | - | 1 | 98 | |||
BC063818.1 | - | 5 | 490 | 6503687 | ||
DA324061.1 | - | 4 | 430 | |||
DA807335.1 | - | 5 | 524 |