Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD3EAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297590.1 | NP_001284519.1 | 3 | 1421 | ||
NM_012099.1 | NP_036231.1 | 3 | 1415 | |||
XM_017026180.1 | XP_016881669.1 | 3 | 1516 | |||
XM_017026181.1 | XP_016881670.1 | 3 | 1586 | |||
GenBank mRNA | AF017633.1 | - | 3 | 939 | ||
AK292183.1 | - | 3 | 980 | |||
BC009705.1 | - | 3 | 972 | |||
BC038992.1 | - | 3 | 943 | 6052122 | ||
BC046141.1 | - | 3 | 1215 | |||
BC065251.1 | - | 3 | 946 | |||
BC108889.2 | - | 3 | 1180 | 40001010 | ||
U86751.1 | - | 3 | 1415 |
Gene Symbol: ERCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166049.1 | NP_001159521.1 | 9 | 1744 | ||
NM_001983.3 | NP_001974.1 | 10 | 1816 | |||
XM_017026460.1 | XP_016881949.1 | 10 | 2217 | |||
XM_017026461.1 | XP_016881950.1 | 11 | 1846 | |||
XM_017026462.1 | XP_016881951.1 | 11 | 1811 | |||
XM_017026463.1 | XP_016881952.1 | 10 | 1803 | |||
XM_017026464.1 | XP_016881953.1 | 10 | 1804 | |||
XM_017026465.1 | XP_016881954.1 | 9 | 2145 | |||
XM_017026466.1 | XP_016881955.1 | 10 | 1779 | |||
GenBank mRNA | AK092039.1 | - | 9 | 1554 |