Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006672.3 | NP_006663.2 | 8 | 1436 | ||
NM_153320.2 | NP_696961.2 | 9 | 1442 | |||
XM_006714970.3 | XP_006715033.1 | 9 | 1451 | |||
XM_006714971.3 | XP_006715034.1 | 8 | 1445 | |||
XM_011514256.2 | XP_011512558.1 | 9 | 1628 | |||
XM_011514257.2 | XP_011512559.1 | 9 | 1619 | |||
XM_011514259.1 | XP_011512561.1 | 9 | 1628 | |||
XM_011514261.2 | XP_011512563.1 | 10 | 1393 | |||
XM_011514263.2 | XP_011512565.1 | 9 | 1195 | |||
XM_017010198.1 | XP_016865687.1 | 9 | 1628 | |||
XM_017010199.1 | XP_016865688.1 | 8 | 1469 | |||
XM_017010200.1 | XP_016865689.1 | 10 | 1433 | |||
XM_017010201.1 | XP_016865690.1 | 10 | 1403 | |||
GenBank mRNA | AF210455.1 | - | 8 | 1337 | ||
AK290796.1 | - | 8 | 1355 | |||
AK290806.1 | - | 8 | 1414 | |||
AK309570.1 | - | 8 | 1407 | |||
AY050498.1 | - | 9 | 1343 | |||
BC033805.1 | - | 9 | 1363 | 5187472 |