Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COPS8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006710.4 | NP_006701.1 | 1 | 584 | ||
NM_198189.2 | NP_937832.1 | 1 | 584 | |||
GenBank mRNA | AI690743.1 | - | 1 | 313 | ||
AK289891.1 | - | 1 | 118 | |||
AK294078.1 | - | 1 | 139 | |||
AK308962.1 | - | 1 | 111 | |||
AK313437.1 | - | 1 | 127 | |||
BC003090.1 | - | 1 | 108 | 3503287 | ||
BC036499.1 | - | 1 | 718 | 5264527 | ||
BC080617.1 | - | 1 | 80 | 6252628 | ||
BF979628.1 | - | 148 | ||||
BG721393.1 | - | 1 | 122 | |||
CR456994.1 | - | 1 | 60 | |||
U51205.1 | - | 1 | 109 |