Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FASTK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258461.1 | NP_001245390.1 | 4 | 830 | ||
NM_006712.4 | NP_006703.1 | 4 | 911 | |||
NM_033015.3 | NP_148936.2 | 3 | 488 | |||
XM_005249932.1 | XP_005249989.1 | 4 | 1103 | |||
XM_005249933.3 | XP_005249990.1 | 4 | 1309 | |||
XM_011515761.2 | XP_011514063.1 | 4 | 911 | |||
XM_011515762.2 | XP_011514064.1 | 4 | 911 | |||
XM_011515763.1 | XP_011514065.1 | 3 | 369 | |||
XM_017011704.1 | XP_016867193.1 | 4 | 1022 | |||
XM_017011705.1 | XP_016867194.1 | 3 | 466 | |||
XM_017011706.1 | XP_016867195.1 | 3 | 658 | |||
GenBank mRNA | AB209783.1 | - | 4 | 731 | ||
AK223132.1 | - | 4 | 821 | |||
AK292232.1 | - | 4 | 796 | |||
AL713798.1 | - | 2 | 1764 | |||
BC000377.2 | - | 4 | 823 | 2820338 | ||
BC006386.2 | - | 3 | 794 | |||
BC011770.2 | - | 4 | 822 | 3831196 | ||
BC039026.2 | - | 3 | 458 | 4940575 | ||
X86779.1 | - | 4 | 815 |