Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB40B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006822.2 | NP_006813.1 | 4 | 447 | ||
XM_005256334.4 | XP_005256391.1 | 4 | 650 | |||
XM_006722271.3 | XP_006722334.1 | 4 | 216 | |||
XM_011523528.2 | XP_011521830.1 | 4 | 569 | |||
XM_017024042.1 | XP_016879531.1 | 5 | 1082 | |||
XM_017024043.1 | XP_016879532.1 | 4 | 559 | |||
GenBank mRNA | AB209711.1 | - | 4 | 425 | ||
AL047483.1 | - | 4 | 425 | |||
BC018039.1 | - | 4 | 447 | 4801271 | ||
U05227.1 | - | 4 | 366 |